Learn More
Potassium bis(oxalato)oxotitanate(IV) dihydrate
Description
Mordant in dyeing, reagent Potassium bis(oxalato)oxotitanate(IV) dihydrate is used as a mordant in dyeing and acts as an analytical reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Odor | Odorless |
| Quantity | 500 g |
| UN Number | UN3288 |
| Solubility Information | Soluble in water. |
| Formula Weight | 354.10 (318.10 Anhydrous) |
| Physical Form | Crystalline |
| Chemical Name or Material | Potassium bis (oxalato)oxotitanate (IV) dihydrate |
Safety and Handling
| Hazard Category | H302+H312 |
| Hazard Statement | GHS H Statement H302-H312 Harmful if swallowed. Harmful in contact with skin. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S. |
| EINECSNumber | 238-475-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.