Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium borohydride, 98%, Thermo Scientific™
Click to view available options
Quantity:
500 g
1 kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics

Specifications
Specifications
Melting Point | >400°C (decomposition) |
Physical Form | Granules |
Quantity | 1 kg |
Linear Formula | KBH4 |
UN Number | UN1870 |
Merck Index | 14,7616 |
Solubility Information | Soluble in water (19g/100g ); methanol (3;9g/100g ); 4:1 water:methanol mixture (13g/100g ). |
IUPAC Name | potassium;boron(1-) |
Formula Weight | 53.94 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H261-H301+H311-H314 |
Hazard Statement | GHS H Statement H261-H301-H311-H331-H314-H318 In contact with water releases flammable gas. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P223-P231+P232-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: POTASSIUM BOROHYDRIDE |
EINECSNumber | 237-360-5 |
RTECSNumber | TS7525000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction