Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium borohydride, 98%, Thermo Scientific™

Catalog No. p-7080224
Click to view available options
Quantity:
500 g
1 kg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13762-51-1
Molecular Formula BH4K
Molecular Weight (g/mol) 53.94
MDL Number MFCD00011396
InChI Key ICRGAIPBTSPUEX-UHFFFAOYSA-N
Synonym potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium
PubChem CID 22892188
SMILES [BH4-].[K+]

Specifications

Melting Point >400°C (decomposition)
Physical Form Granules
Quantity 500 g
Linear Formula KBH4
UN Number UN1870
Merck Index 14,7616
Solubility Information Soluble in water (19g/100g ); methanol (3;9g/100g ); 4:1 water:methanol mixture (13g/100g ).
IUPAC Name potassium boranuide
Formula Weight 53.94
Percent Purity 98%
Odor Odorless
Refractive Index 1.494
Sensitivity Moisture sensitive
Density 1.17 g/mL
Chemical Name or Material Potassium borohydride
Show More Show Less
Hazard Category H261-H301+H311-H314
Hazard Statement GHS H Statement
H261-H301-H311-H331-H314-H318
In contact with water releases flammable gas.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: POTASSIUM BOROHYDRIDE
EINECSNumber 237-360-5
RTECSNumber TS7525000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.