Learn More
Potassium borohydride, 98%
Description
Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plasticsPotassium borohydride is used as a selective reducing agent for ketones, aldehydes and acid chlorides. It is used as a precursor in the production of chloramphenicol, vitamin A, thiopenicol, atropine and scopolamine. It is used to prepare high dispersive copper nanoparticles by the chemical reduction method using potassium borohydride as a reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >400°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Assay Percent Range | 98% |
Solubility Information | Soluble in water (19g/100g ). methanol (3.9g/100g ). 4:1 water:methanol mixture (13g/100g ).Soluble in methanol. Insoluble in ethanol,ether and tetrahydrofuran. |
IUPAC Name | potassium boranuide |
Formula Weight | 53.94 |
Odor | Odorless |
Refractive Index | 1.494 |
Show More |
Safety and Handling
Hazard Category | H261-H301+H311-H314 |
Precautionary Statement | P223-P231+P232-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | DOT Class: 4.3; Packing Group: I |
EINECSNumber | 237-360-5 |
RTECSNumber | TS7525000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.