Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Carbonate, 1.5-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Specifications
Physical Form | Crystals |
Quantity | 500 g |
Formula Weight | 165.24 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.04kg |
Chemical Name or Material | Potassium Carbonate, 1.5-Hydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction