Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium carbonate sesquihydrate, 98.5%, ACS reagent

Catalog No. p-3777018
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC347202500 250 g Glass bottle
AC347200010 1 kg Glass bottle
AC347200025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC347202500 Supplier Thermo Scientific Chemicals Supplier No. 347202500
Only null left
Add to Cart

CAS: 6381-79-9 | C2H6K4O9 | 330.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-79-9
Molecular Formula C2H6K4O9
Molecular Weight (g/mol) 330.45
MDL Number MFCD00149925
InChI Key WWVGGBSGYNQKCY-UHFFFAOYSA-J
IUPAC Name tetrapotassium trihydrate dicarbonate
SMILES O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point 891.0°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 98.5%
Merck Index 15, 7740
Solubility Information Solubility in water: soluble in water., insoluble in alcohol
Formula Weight 165.24
Percent Purity 98.5%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Potassium carbonate sesquihydrate, For analysis ACS
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.