Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Channel Kv2.2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$382.00 - $610.00

Specifications

Antigen Potassium Channel Kv2.2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP257427
SDP
View Documents
Novus Biologicals
NBP257427
100 μL
Each for $610.00
Only null left
Add to Cart
 
NB393971
SDP
View Documents
Novus Biologicals
NBP25742725UL
25 μL
Each for $382.00
Only null left
Add to Cart
 
Description

Description

Potassium Channel Kv2.2 Polyclonal specifically detects Potassium Channel Kv2.2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Potassium Channel Kv2.2
Polyclonal
Rabbit
Autophagy, Neuronal Cell Markers, Neuroscience, Neurotransmission, Potassium Channels
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
9312
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QHISTILLEETPSQGDRPLLGTEVSAPCQGPSKGLSPRFPKQKLFPFSSRERRSFTEIDTGDDEDFLELP
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human
delayed rectifier potassium channel protein, KV2.2, potassium channel Kv2.2, potassium voltage-gated channel subfamily B member 2, potassium voltage-gated channel, Shab-related subfamily, member 2, voltage-gated potassium channel subunit Kv2.2
KCNB2
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.