Learn More
Description
Specifications
Specifications
| Antigen | Potassium Channel Kv3.1 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | P48547 |
| Gene Alias | FLJ41162, FLJ42249, FLJ43491, KV3.1, KV4, MGC129855, NGK2, potassium voltage-gated channel subfamily C member 1, potassium voltage-gated channel, Shaw-related subfamily, member 1, voltage-gated potassium channel protein KV3.1, Voltage-gated potassium channel subunit Kv3.1, Voltage-gated potassium channel subunit Kv4 |
| Gene Symbols | KCNC1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
