Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Channel Kv3.1, p Ser503 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB300279
Description
Potassium Channel Kv3.1 Polyclonal specifically detects Potassium Channel Kv3.1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Potassium Channel Kv3.1 (p Ser503) | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 1:1000, Immunohistochemistry-Frozen 1:1000 | |
P25122 | |
KCNC1 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser503 conjugated to KLH. Accession # P25122 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼100k Kv3.1 voltage-gated potassium channel protein phosphorylated at Ser503. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
FLJ41162, FLJ42249, FLJ43491, KV3.1, KV4, MGC129855, NGK2, potassium voltage-gated channel subfamily C member 1, potassium voltage-gated channel, Shaw-related subfamily, member 1, voltage-gated potassium channel protein KV3.1, Voltage-gated potassium channel subunit Kv3.1, Voltage-gated potassium channel subunit Kv4 | |
Rabbit | |
100 kDa | |
0.1 mL | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Potassium Channels | |
3746 | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction