Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Chlorate
SCI_ED

Catalog No. p-4416424 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade

Catalog No. S25482

Only null left
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
Synonym potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
PubChem CID 6426889
IUPAC Name potassium;chlorate
SMILES [O-]Cl(=O)=O.[K+]

Specifications

Color White
Quantity 100 g
Assay Percent Range >95%
Linear Formula KClO3
Formula Weight 122.55
Grade Laboratory
Packaging Lab Grade
Chemical Name or Material Potassium Chlorate
ShelfLife 5 years
DOTInformation DOT Class 5.1, II, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.