Learn More
Potassium chlorate, 99+%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 356.0°C to 368.0°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 2.5 kg |
| Assay Percent Range | 99+% |
| Linear Formula | KClO3 |
| Fieser | 01,927 |
| Merck Index | 15, 7741 |
| Solubility Information | Solubility in water: 73g/L (20°C). Other solubilities: 555g/L hot water (100°C),soluble in alkalies,1g/50mL glycerol,practically insoluble in alcohol,insoluble in ether |
| Formula Weight | 122.55 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled. May cause fire or explosion; strong oxidizer. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the ris |
| EINECSNumber | 223-289-7 |
| RTECSNumber | FO0350000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.