Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, 3 Molar Electrode Filling Solution, Reagents

Catalog No. 01334266
Change view
Click to view available options
Quantity:
120 mL
250 mL
500 mL
1 L
4 L
10 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-266 1 L
01-334-268 10 L
01-334-267 120 mL
01-334-265 250 mL
01-334-264 4 L
01-334-263 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-334-266 Supplier Reagents Holdings Llc Supplier No. CP1830001A
Only null left
Add to Cart
Add to Cart

Potassium Chloride, 3 Molar Electrode Filling Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Chloride, 3 Molar Electrode Filling Solution
Boiling Point 103°C
Color Colorless
CAS 7732-18-5, 7447-40-7
pH 5.4 to 8.6
Physical Form Liquid
Quantity 1 L
Molecular Formula KCl/H2O
Synonym Muriate of Potash
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1129
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.