Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, 3 M, saturated with Silver Chloride, Electrode Filling Solution, Ricca Chemical
SDP

Catalog No. 591832 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 7447-40-7, 7732-18-5, 7761-88-8
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Color Colorless
CAS Min % 17.89, 80.0, 0.0034
CAS Max % 18.62, 83.0, 0.0036
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼7
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 3Molar,saturated with Silver Chloride
Chemical Name or Material Potassium Chloride, Electrode Filling Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.