Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chloride, 3M aq. soln.

Catalog No. p-4824888
Click to view available options
Quantity:
250 mL
1 L

For Refilling Calomel Electrodes | CAS: 7447-40-7 | ClK | 74.55 g/mol

For refilling calomel electrodesPotassium chloride, 3M aq. solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7, 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 250 mL
Assay Percent Range 3M
Solubility Information Miscible with water.
Formula Weight 74.55
Concentration or Composition (by Analyte or Components) Potassium chloride: 23%; Water: 77%
Odor Odorless
Vapor Pressure 17mm Hg at 20°C
Chemical Name or Material Potassium chloride
EINECSNumber 231-211-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.