Potassium chloride, 99.997% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1083918

Quantity 50g

 View more versions of this product

Catalog No. AA1083918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7447-40-7
    Physical Form Powder/Crystalline/Beads
    Assay Percent Range 99.997% (metals basis)
    Molecular Formula KCl
    Formula Weight 74.55
    MDL Number MFCD00011360
    Chemical Name or Material Potassium chloride

    Specifications

    Physical Form Powder/Crystalline/Beads
    Formula Weight 74.55
    Quantity 50g
    Color White
    EINECS Number 231-211-8
    Odor Odorless
    Assay Percent Range 99.997% (metals basis)
    Molecular Formula KCl
    Appearance White Powder/crystalline/beads
    MDL Number MFCD00011360
    CAS 7447-40-7
    Health Hazard 1 H303: May be harmful if swallowed.
    Chemical Name or Material Potassium chloride

    H303: May be harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit