Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chloride, 99.999%, (trace metal basis)

Catalog No. p-4229013
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle

CAS: 7447-40-7 | ClK | 74.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420.0°C
Melting Point 770.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99.999%
Linear Formula KCl
Fieser 06,48
Merck Index 15,7742
Solubility Information Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone
IUPAC Name potassium chloride
Formula Weight 74.54
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Potassium chloride
Show More Show Less
EINECSNumber 231-211-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.