Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron trichloride, 1M solution in hexane, AcroSeal™

Catalog No. AC198928000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 10294-34-5 | BCl3 | 117.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boron trichloride
Color Colorless to Yellow
Name Note 1M Solution in Hexane
CAS 10294-34-5, 110-54-3
Molecular Formula BCl3
Linear Formula BCl3
MDL Number MFCD00011313
Concentration 0.9 to 1.1M
Fieser 01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44
Density 0.7380g/mL
Synonym boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
Solubility Information Solubility in water: may decompose.
InChI Key FAQYAMRNWDIXMY-UHFFFAOYSA-N
SMILES ClB(Cl)Cl
IUPAC Name trichloroborane
Molecular Weight (g/mol) 117.16
PubChem CID 25135
Formula Weight 117.17
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.738
Quantity 800 mL
Flash Point −17°C
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.