Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Chloride Conductivity Standard, 1000 μS/cm at 25°C, Ricca Chemical

Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cube
Cubitainer
Natural Poly Bottle
Description
- Tested in ISO 17025 Accredited Laboratories.
- Manufactured in cGMP compliant, FDA registered facilities.
- Certified Traceable to NIST Standard Reference Material when available.
- Many C of As and labels contain TDS and conductance information.
- Tightest Specifications in the industry, lowest lot-to-lot variability.
Specifications
Specifications
Packaging | Natural Poly Bottle |
CAS Min % | 98.0%, 0.05% |
CAS Max % | 100.0%, 0.05% |
Physical Form | Liquid |
Color | Colorless |
pH | Approximately 7 |
Boiling Point | 100°C |
Solution Type | Conductivity Standard |
Type | Conductivity Standard |
Quantity | 500 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction