Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Chloride Conductivity Standard, 50 μS/cm at 25°C, Ricca Chemical

Click to view available options
Quantity:
250 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
Packaging | Natural Poly Bottle |
CAS Min % | 98.0, 0.0025 |
CAS Max % | 100.0, 0.0026 |
Physical Form | Liquid |
Color | Colorless |
pH | Approximately 7 |
Boiling Point | 100°C |
Solution Type | Conductivity Standard |
Type | Conductivity Standard |
Quantity | 250 mL |
Show More |
Safety and Handling
ShelfLife | 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction