Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Chloride, Crystal, Multi-Compendial, U.S.P., J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Specifications
Physical Form | Crystals |
Quantity | 2.5 kg |
IUPAC Name | potassium chloride |
Formula Weight | 74.55 |
Grade | Multi-Compendial/USP |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.99kg |
Chemical Name or Material | Potassium Chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction