Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride (Crystalline/Certified ACS), Fisher Chemical PROMO

Catalog No. P217500FB25 Shop All Fisherbrand Products
Click to view available options
:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Potassium chloride for calomel cells.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55 g/mol
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420°C
Melting Point 770°C
Color White
pH 6
Physical Form Solid
Quantity 500 g
Assay Percent Range 99.0 to 100.5%
Formula Weight 74.55 amu
Percent Purity 99.0 to 100.5%
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Potassium Chloride
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement

Emergency Overview: May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1

Flammability:1

Instability:0

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.