Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Chloride, Granular
SCI_ED

Catalog No. p-4416425 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Color White
Quantity 100 g
Assay Percent Range >95%
Linear Formula KCl
Formula Weight 74.56
Packaging Reagent Grade
Density 1.987g/mL
Chemical Name or Material Potassium Chloride
ShelfLife 2 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.