Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Chloride, Granular, BiotechGrade, 99-100.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Potassium Chloride, Granular, BiotechGrade is commonly used as a laboratory reagent as a standard. It may be used as a calibration standard ionic solution for measuring electrical conductivity because potassium chloride solutions, when prepared carefully, produce repeatable measurable properties. Potassium chloride is also a good source of ionic chloride, which should precipitate insoluble chloride salts upon addition to a solution of an appropriate metal ion.
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 12 kg |
| Percent Purity | 99 to 100.5% |
| Grade | Biotech |
| Loss on Drying | 0.01 |
| Odor | Odorless |
| Packaging | Poly Pail |
| Appearance | Clear |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction