Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Granular, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle

PO170, 7447-40-7, KCl

Spectrum™ Potassium Chloride, Granular, USP is used to treat hypokalemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade USP
CAS 7447-40-7
CAS Max % 1
Molecular Formula ClK
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
Percent Purity 99 to 100.5%
Bromide 0.001
Calcium and Magnesium To pass test
Iodide 5e-005
Loss on Drying 0.01
Packaging Poly Bottle
Quantity 500 g
Show More Show Less