Alfa Aesar™ Potassium chloride, Optical Grade

Manufacturer: Alfa Aesar™ 2210914

Quantity 25g

 View more versions of this product

Catalog No. AA2210914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7447-40-7
    Formula Weight 74.55
    MDL Number MFCD00011360
    Merck Index 14,7621
    Chemical Name or Material Potassium chloride
    Grade Optical Grade

    Specifications

    Formula Weight 74.55
    RTECS Number TS8050000
    Odor Odorless
    Physical Form Broken Crystals
    Refractive Index 1.490
    MDL Number MFCD00011360
    Merck Index 14,7621
    Boiling Point 1500°C (subl.)
    Melting Point 773°C
    TSCA TSCA
    Sensitivity Hygroscopic
    CAS 7447-40-7
    Quantity 25g
    Density 1.984g/mL
    Chemical Name or Material Potassium chloride
    Grade Optical Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit