Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Chloride Solution, For Ag/AgCl electrodes, ∼3 M KCl, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
250 mL
1 L
Description
KCl exists as a white ionic solid, and can be prepared by neutralizing HCl with potassium hydroxide solution. It is less soluble in hot water as compared to cold water. It occurs naturally as the minerals carnallite and sylvine. It is generally used as a fertilizer.

Specifications
Specifications
CAS | 7447-40-7 |
Quantity | 250 mL |
Linear Formula | KCl |
Synonym | Potassium chloride solution saturated with AgCl |
Formula Weight | 74.55 |
Density | 1.13 g/mL (at 20°C) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction