Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chlorochromate, 97+%, Thermo Scientific™

Catalog No. AAA1900014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1900014 25 g
AAA1900022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AAA1900014 Supplier Thermo Scientific Chemicals Supplier No. A1900014
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16037-50-6
Molecular Formula ClCrKO3
Molecular Weight (g/mol) 174.541
MDL Number MFCD00064578
InChI Key PEBPTQFCMJWPGZ-UHFFFAOYSA-M
Synonym potassium chlorotrioxochromate, chlorotrioxochromic acid, potassium chlorochromate, chromate 1-, chlorotrioxo-, potassium, t-4, chromate 1-, chlorotrioxo-, potassium 1:1 , t-4
PubChem CID 23689123
IUPAC Name potassium;trioxochromium;chloride
SMILES O=[Cr](=O)=O.[Cl-].[K+]

Specifications

Melting Point 290°C to 292°C
Quantity 25 g
Linear Formula KClCrO3
UN Number UN1479
Formula Weight 174.55
Percent Purity ≥97%
Odor Odorless
Chemical Name or Material Potassium chlorochromate
Hazard Category Danger
Hazard Statement GHS H Statement
H272-H350-H317
May intensify fire; oxidizer.
May cause cancer.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P221-P273-P201-P309-P310-P501a
Take any precaution to avoid mixing with combustibles.
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.