Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloroplatinate, 0.2% (w/v) in 10% (v/v) Hydrochloric Acid, Ricca Chemical
SDP

Catalog No. 595016
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 16921-30-5, 7732-18-5, 7647-01-0
Molecular Formula Cl6K2Pt
Molecular Weight (g/mol) 485.98
MDL Number MFCD00011389
InChI Key DPAIVKJGTXERIM-UHFFFAOYSA-H
Synonym potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride
PubChem CID 61856
IUPAC Name dipotassium hexachloroplatinumdiuide
SMILES [K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Color Yellow
CAS Min % 0.18, 87.0, 10.32
CAS Max % 0.18, 91.0, 10.75
Percent Purity 0.2%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix 10% (v/v) Hydrochloric Acid
pH <1
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.2% (w/v)
Chemical Name or Material Potassium Chloroplatinate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.