Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Chromate, 5% (w/v), Chloride Free, Indicator for Argentometric Titrations, Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
| CAS Min % | 4.67, 93.0, 7.0E-4 |
| CAS Max % | 4.86, 97.0, 7.0E-4 |
| Percent Purity | 5% |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 5% (w/v) Aqueous Solution |
| Packaging | Natural Poly Bottle |
| pH | Alkaline |
| Quantity | 4 L |
| Chemical Name or Material | Potassium Chromate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction