Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Potassium chromate, ACS, 99.0% min

Catalog No. p-4407729
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For Use in Finishing Leather, Rust Proofing of Metals, Textile Mordant | CAS: 7789-00-6 | CrK2O4 | 194.19 g/mol

In finishing of leather, rust proofing of metals, textile mordant, as an analytical lab reagent. Potassium chromate enjoys a broad assortment of industrial purposes. In the fabric industry, manufacturers use potassium chromate to tan leather and dye clothing items and materials. In science, potassium chromate is often used as an indicator, meaning it can identify the presence, location and quantity of certain elements such as iron. Beyond these applications, potassium chromate also functions as an oxidizing agent in manufacturing and scientific experiments. Some nuclear power plants currently use substances other than potassium chromate to inhibit corrosion in closed cooling-water systems. Potassium chromate is applied as a chemical indicator of chlorine ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
IUPAC Name dipotassium;dioxido(dioxo)chromium
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Melting Point 975°C
Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.0% min.
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 194.19
Grade ACS
Odor Odorless
Appearance Yellow crystals
Vapor Pressure 0 hPa
Density 2.73 g/cm3 at 20°C
Chemical Name or Material Potassium chromate, For analysis ACS
Show More Show Less
Hazard Category H315-H317-H319-H335-H340-H350i-H373
Precautionary Statement P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
DOTInformation DOT Class: 9; Packaging Group: III
EINECSNumber 232-140-5
RTECSNumber GB2940000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.