Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium chromate, ACS reagent, meets the requirements of Reag.Ph.Eur.
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 975.0°C |
| Quantity | 100 g |
| Linear Formula | K2CrO4 |
| Merck Index | 15,7743 |
| Solubility Information | Solubility in water: 640g/L (20°C). Other solubilities: insoluble in alcohol |
| Formula Weight | 194.2 |
| Grade | ACS Reagent |
| Packaging | Glass bottle |
| Chemical Name or Material | Potassium chromate |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. Causes skin irritation. May cause respiratory irritation. May cause cancer by inhalation. May cause genetic defects. Causes serious eye irritation. Very toxic |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at r |
| EINECSNumber | 232-140-5 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction