Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chromate TS, 10% (w/v), Chloride Free, Indicator for Argentometric Titrations, Ricca Chemical
SDP

Catalog No. 600516 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle

Catalog No. 6005-16

Supplier: Ricca Chemical Company 600516

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7789-00-6, 7732-18-5, 7761-88-8
Molecular Formula CrK2O4
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
IUPAC Name dipotassium dioxochromiumbis(olate)
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

CAS Min % 8.91, 89.0, 7.0E-4
CAS Max % 9.27, 93.0, 7.0E-4
Concentration 10% (w/v)
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Yellow
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Potassium Chromate, Indicator for Argentometric Titrations, Test Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.