Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Citrate Monohydrate (Granular/USP/FCC), Fisher Chemical™

Catalog No. p-187054 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle

Chemical Identifiers

CAS 1534146
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
MDL Number MFCD00150442
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
PubChem CID 2735208
ChEBI CHEBI:64746
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]

Specifications

Alkalinity Pass Test (FCC/USP)
Melting Point 180°C
Color White
pH 7.5 to 9.0
Packaging Poly Bottle
Linear Formula KO2CCH2C(OH)(CO2K)CH2CO2K·H2O
Quantity 500 g
Residual Solvents Pass Test (USP)
Formula Weight 324.41
Percent Purity 99.0 to 100.5% (FCC)
Grade FCC/USP
Physical Form Solid
Chemical Name or Material Potassium Citrate Monohydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.