Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper(II) acetate monohydrate, 98+%, extra pure
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent

Specifications
Specifications
Boiling Point | 240.0°C |
Melting Point | 115.0°C |
Color | Blue to Blue-Green |
Physical Form | Crystalline Powder |
Quantity | 2.5 kg |
Assay Percent Range | 98% min. (Iodometry) |
Linear Formula | (CH3CO2)2Cu·H2O |
Merck Index | 15,2614 |
Solubility Information | Solubility in water: 72g/L (20°C). Other solubilities: soluble in alcohol and ether |
Formula Weight | 199.65 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with wat |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction