Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium cyanide, 97+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
500g
Packaging:
Plastic bottle
151-50-8
CKN
65.12
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
9032
potassium;cyanide
[C-]#N.[K+]
151-50-8
CKN
65.12
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
9032
potassium;cyanide
[C-]#N.[K+]
C-X bond formation reagent
Assay Percent Range 97% min. (Argentometry)
Linear Formula CKN
Fieser 05,553; 07,299; 08,409; 10,324; 11,433; 13,87; 16,22
Merck Index 15, 7747
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: ethanol 4.5g/L (19.5°C)
Formula Weight 65.12
Physical Form Powder Solid
Percent Purity 97+%
Grade Analysis
Chloride 500ppm max.
Chlorine (Cl) 500ppm max.
Iron (Fe) 100ppm max.
Lead (Pb) 5ppm max.
Packaging Plastic bottle
Phosphate 50ppm max.
Sodium (Na) 5000ppm max.
Solubility Solubility in water: 400g/L (20°C). Other solubilities: ethanol 4.5g/L (19.5°C)
Sulfate 200ppm max.
Sulfur (S) 10ppm max.
Thiocyanate 100ppm max.
Boiling Point 1625.0°C
Color Colorless
Melting Point 634.0°C
Quantity 250g
Chemical Name or Material Potassium cyanide, p.a.
Show More Show Less

GHS H Statement
May be corrosive to metals.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with

GHS P Statement
Absorb spillage to prevent material damage.
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at r

GHS Signal Word: Danger

EINECSNumber : 205-792-3

RTECSNumber : TS8750000

TSCA : TSCA