Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium cyanide, 98+%, for analysis, Thermo Scientific Chemicals

Catalog No. AC196605000
Click to view available options
Quantity:
250 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.12
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032

Specifications

Boiling Point 1625.0°C
Melting Point 634.0°C
Color Colorless
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range 98% min. (Argentometry)
Linear Formula CKN
Fieser 05,553; 07,299; 08,409; 10,324; 11,433; 13,87; 16,22
Merck Index 15, 7747
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: ethanol 4.5g/L (19.5°C)
SMILES [C-]#N.[K+]
IUPAC Name potassium;cyanide
Formula Weight 65.12
Percent Purity 98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Potassium cyanide, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with
Precautionary Statement GHS P Statement
Absorb spillage to prevent material damage.
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at r
EINECSNumber 205-792-3
RTECSNumber TS8750000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.