Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium cyanide, ACS, 96.0% min, Thermo Scientific Chemicals

Catalog No. p-4412858
Click to view available options
Quantity:
50 g
250 g
1 kg

KCN is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von Richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Boiling Point 1625°C
Melting Point 634°C
Physical Form Granules
Quantity 50 g
Assay Percent Range 96.0% min.
Beilstein 4652394
Solubility Information Highly soluble in water. Soluble in methanol,glycerol,and formamide. Slightly soluble in ethanol.
Formula Weight 65.12
Grade ACS Reagent
Sensitivity Hygroscopic
Density 1.52
Chemical Name or Material Potassium Cyanide, For analysis ACS
Show More Show Less
Hazard Category H290-H300+H310+H330-H372
Precautionary Statement P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P390-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 205-792-3
RTECSNumber TS8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.