Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, 0.100 Normal (N/10), Reagents

Catalog No. 01336450
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Catalog No. Quantity
01-336-450 10 L
01-336-449 1 L
01-336-448 20 L
01-336-447 4 L
4 options

Catalog No. 01-336-450

Supplier: Reagents Holdings Llc C728253010F

Only null left
Add to Cart
Add to Cart

Potassium Dichromate, 0.100 Normal (N/10), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7778-50-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Potassium bichromate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Orange
Physical Form Liquid
Quantity 10 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.0036
Chemical Name or Material Potassium Dichromate, 0.100 Normal (N/10)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.