Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Dichromate, ACS, Reagents
Click to view available options
Quantity:
500 g
3 kg
Specifications
Specifications
Boiling Point | 500°C |
Melting Point | 398°C |
Color | Orange/Red |
Physical Form | Solid |
Quantity | 3 kg |
Grade | Pure |
Packaging | Poly Bottle |
Specific Gravity | 2.676 |
Chemical Name or Material | Potassium Dichromate, ACS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction