Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Ethoxide

Catalog No. 333379500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
33-337-9500G 500 g
33-337-9100G 100 g
2 options
Danger

Catalog No. 33-337-9500G

Supplier: Sigma Aldrich 333379500G

Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium ethoxide can be used:As a catalyst for transesterification reactions for the synthesis of biodiesel.To synthesize nitrogen doped carbon nano-sheets to be used as an anode in sodium ion batteries.To promote the arylation reaction of unactivated arenes to synthesize biaryl compounds.As a precursor for the synthesis of thin films of (Na0.5K0.5)NbO3 (NKN) and K3Li2Nb5O15 (KLN).For ethoxylation of p-chloronitrobenzene in the presence of phase transfer catalysts.

Specifications

CAS 917-58-8
Molecular Formula C2H5KO
Linear Formula KOC2H5
MDL Number MFCD00054277
Quantity 500 g
Molecular Weight (g/mol) 84.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.