Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium ethyl xanthate, 97+%

Catalog No. aaa1145022
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 140-89-6 | C3H5KOS2 | 160.29 g/mol

Potassium ethyl xanthate is used as a reagent for extraction of metal ions into organic solvents and in the preparation of 2-mercapto benzimidazoles and 2-mercapto benzoxazoles by reacting with o-phenylenediamines and o-amino phenols respectively. It acts as a precursor in the production of unsymmetrical sulfides through cross-coupling reaction and as a floatation agent in the mining industry. It is involved in the determination of cadmium in water samples by substoichiometric radiochemical method and the adsorption of colloidal dye disperse Blue 3 onto pretreated polyester fabric.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium ethyl xanthate is used as a reagent for extraction of metal ions into organic solvents and in the preparation of 2-mercapto benzimidazoles and 2-mercapto benzoxazoles by reacting with o-phenylenediamines and o-amino phenols respectively. It acts as a precursor in the production of unsymmetrical sulfides through cross-coupling reaction and as a floatation agent in the mining industry. It is involved in the determination of cadmium in water samples by substoichiometric radiochemical method and the adsorption of colloidal dye disperse Blue 3 onto pretreated polyester fabric.

Solubility
Soluble in water.

Notes
Light sensitive. Incompatible with strong acids, strong bases and strong oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-89-6
Molecular Formula C3H5KOS2
Molecular Weight (g/mol) 160.29
MDL Number MFCD00004931
InChI Key JCBJVAJGLKENNC-UHFFFAOYSA-M
Synonym potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide
PubChem CID 2735045
IUPAC Name potassium;ethoxymethanedithioate
SMILES [K+].CCOC([S-])=S

Specifications

Melting Point ∽205°C (decomposition)
Density 1.558 g/mL
Flash Point 205°C (401°F)
Assay Percent Range 98%
Quantity 100 g
Beilstein 3596974
Merck Index 14,7696
Solubility Information Soluble in water.
Formula Weight 160.3
Percent Purity 98%
Chemical Name or Material Potassium ethyl xanthate
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 205-439-3
RTECSNumber FG1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.