Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Ferricyanide, 10% (w/v), Ricca Chemical

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Poly Bottle
Specifications
Specifications
Color | Orange-Yellow |
CAS Min % | 89.0, 8.91 |
CAS Max % | 93.0, 9.28 |
Percent Purity | 10% |
Grade | Laboratory |
Packaging | Amber Poly Bottle |
Solvent or Matrix | Aqueous Solution |
Quantity | 500 mL |
Physical Form | Liquid |
Concentration or Composition Notes | 10% (w/v) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction