Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Ferricyanide, ACS Reagent (Potassium Hexacyanoferrate (III)), Reagents

Catalog No. 01337847
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg

Potassium Ferricyanide, ACS Reagent (Potassium Hexacyanoferrate (III)), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym Potassium Hexacyanoferrate III, Red Prussiate, Potassium Ferricyanate
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 300°C
Color Orange/Red
Physical Form Solid
Quantity 25 kg
Grade ACS
Packaging Fiber Drum
Specific Gravity 1.89
Chemical Name or Material Potassium Ferricyanide, ACS Reagent (Potassium Hexacyanoferrate (III))
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.