Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium ferrocyanide trihydrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Plastic bottle
14459-95-1
C6FeK4N6·3H2O
422.4
MFCD00167023
Potassium hexacyanoferrate(II)
14459-95-1
C6FeK4N6·3H2O
422.4
MFCD00167023
Potassium hexacyanoferrate(II)
Assay Percent Range 99+%
Linear Formula K4Fe(CN)6·3H2O
Merck Index 15, 7752
Solubility Information Solubility in water: 270g/L (12°C). Other solubilities: insoluble in alcohol
Formula Weight 422.4
Physical Form Crystalline Powder
Percent Purity 99+%
Grade Analysis
Chloride 100ppm max
Chlorine (Cl) 100ppm max.
Density 1.8500g/mL
Insoluble Matter 0.005% max
Packaging Plastic bottle
Solubility Solubility in water: 270g/L (12°C). Other solubilities: insoluble in alcohol
Specific Gravity 1.85
Sulfate 100ppm max.
Color Yellow
Melting Point 70.0°C
Quantity 500g
Chemical Name or Material Potassium ferrocyanide trihydrate, p.a.
Show More Show Less

GHS H Statement:
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE