Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium ferrocyanide trihydrate, 99+%, for analysis

Catalog No. p-43113
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 14459-95-1 | C6H6FeK4N6O3 | 422.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
Synonym Potassium hexacyanoferrate(II)
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 70.0°C
Color Yellow
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99+%
Linear Formula K4Fe(CN)6·3H2O
Merck Index 15, 7752
Solubility Information Solubility in water: 270g/L (12°C). Other solubilities: insoluble in alcohol
Formula Weight 422.4
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.85
Density 1.8500g/mL
Chemical Name or Material Potassium ferrocyanide trihydrate, p.a.
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.