Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Ferrocyanide Trihydrate (Potassium Hexacyanoferrate), Reagents

Catalog No. 01337107
Click to view available options
Quantity:
100 g
500 g

Potassium Ferrocyanide Trihydrate (Potassium Hexacyanoferrate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
Synonym Yellow Prussiate, Potassium Hexacyanoferrate (II) Trihydrate, Tetrapotassium iron (II) hexacyanide trihydrate
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 70°C
Color Pale Yellow
pH 9.5
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.85
Chemical Name or Material Potassium Ferrocyanide Trihydrate (Potassium Hexacyanoferrate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.