Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, 50% (w/v), Neutralized, Ricca Chemical
SDP

Catalog No. 610132 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
610132 1 L Natural Poly Bottle
610116 500 mL Natural Poly Bottle
61011 4 L Natural Poly Bottle
61012-5 10 L Cubitainer
61015 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 610132 Supplier Ricca Chemical Company Supplier No. 610132
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7789-23-3 , 7732-18-5
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Color Colorless to Pink
CAS Min % 65.0, 32.68
CAS Max % 68.0, 34.01
Percent Purity 50%
Grade Laboratory
Packaging Natural Poly Bottle
Treatment(s) Neutralized
Solvent or Matrix Aqueous Solution
pH ∼7
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 50% (w/v)
Chemical Name or Material Potassium Fluoride, Neutralized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.