Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Fluoride, 50% (w/w), pH 8.3, Ricca Chemical

Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
CAS Min % | 70.0, 49.0 |
CAS Max % | 73.0, 51.0 |
Percent Purity | 50% |
Grade | Laboratory |
Concentration or Composition (by Analyte or Components) | 50% (w/w) Aqueous Solution |
Packaging | Cubitainer |
pH | 8.3 |
Quantity | 20 L |
Chemical Name or Material | Potassium Fluoride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction