Learn More
Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Potassium fluoride is used in organic chemistry as a fluorinating agent to replace other halide ions like chorine, bromine etc. It is used as a catalyst in the manufacture of polyurethanes. Further, it is widely used for etching of glass, in disinfectant formulations and manufacture of optical glass. It is used in metal finishing, batteries, coatings and photographic chemicals. It is utilized for the study of ion-specific swelling and de-swelling of ampholytic polymer gels as well as in the measurement of electronic polarizabilities of ions in polymers of alkali halides. It finds application in the electronic industry as a metal surface treatment product. It is used as a preservative, a food additive, a catalyzer and a water absorbing agent. In the Finkelstein reaction, it is actively involved in the conversion of chlorocarbons to fluorocarbons using polar solvents like dimethyl formamide and ethylene glycol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1505°C |
Melting Point | 859°C |
Physical Form | Crystalline |
Quantity | 50 g |
Assay Percent Range | 99.99% (metals basis) |
Beilstein | 3902818 |
Solubility Information | Freely soluble in water. Insoluble in alcohol. |
IUPAC Name | potassium;fluoride |
Formula Weight | 58.1 |
Refractive Index | 1.363 |
Show More |
Safety and Handling
Hazard Category | H301+H311+H331-H318 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 232-151-5 |
RTECSNumber | TT0700000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.