Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium fluoride, anhydrous, 99%, Thermo Scientific Chemicals

Catalog No. AA1413030
Click to view available options
Quantity:
50 g
250 g
1 kg

Potassium fluoride is used in metal finishing, batteries, coatings and photographic chemicals. It is utilized for the study of ion-specific swelling and de-swelling of ampholytic polymer gels as well as in the measurement of electronic polarizabilities of ions in polymers of alkali halides. It finds application in the electronic industry as a metal surface treatment product. It is used as a preservative, a food additive, a catalyzer and a water absorbing agent. In the Finkelstein reaction, it is actively involved in the conversion of chlorocarbons to fluorocarbons using polar solvents like dimethyl formamide and ethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
SMILES [F-].[K+]

Specifications

Boiling Point 1505°C
Melting Point 859°C
Color White
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 99%
Beilstein 3902818
Solubility Information Soluble in water and aqueous hydrogen fluoride. Insoluble in alcohol.
IUPAC Name potassium;fluoride
Formula Weight 58.1
Odor Odorless
Refractive Index 1.363
Sensitivity Hygroscopic
Appearance White various forms (powder/flake/crystalline/beads, etc.)
Vapor Pressure 0.13 hPa at 752°C
Density 2.48 g/cm3
Chemical Name or Material Potassium fluoride, anhydrous
Show More Show Less
Hazard Category H301+H311+H331-H318
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1, Packing Group: III
EINECSNumber 232-151-5
RTECSNumber TT0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.